Methyl calcein blue hydrate indicator for complexometry copper

Toimittaja: TCI
Varoitus

M0825-1G
TCIAM0825-1GEA 108 EUR
TCIAM0825-1G
Methyl calcein blue hydrate indicator for complexometry copper
Methyl calcein blue hydrate
Kaava: C₁₄H₁₅NO₅
MDL numero: MFCD00167005
CAS-numero: 81028-96-8
EINECS: 279-666-1

Order Now

Testitulosten erittely

Appearance Light yellow to Yellow to Green powder to crystal


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR