MOPS ≥98.0% (by titrimetric analysis) Good's buffer component for biological research

Toimittaja: TCI
Varoitus

M0707-25G M0707-250G
TCIAM0707-25GEA 17.6 EUR
TCIAM0707-25G TCIAM0707-250G
MOPS ≥98.0% (by titrimetric analysis) Good's buffer component for biological research
MOPS
Kaava: C₇H₁₅NO₄S
Sulamispiste: > 250 °C
Leimahduspiste: (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006183
CAS-numero: 1132-61-2
EINECS: 214-478-5
Merck Index: 14,06265
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43cc8969484da0d2bb96720fbe89511b79505dbf&vid=7cba1a1fd84aac7a09bb1ebc0615dfb97fce6bc6&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Absorbance max. 0.15 (0.1mol/L, H2O, 260nm)
Solubility in Water almost transparency
Heavy metals (as Pb) max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR