Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥95.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

Synonyms: Molybdenum(VI)dioxy Acetylacetonate, Acetylacetone Molybdenum(VI)dioxy Salt, Molybdenum(VI) oxide bis(acetylacetonate), Molybdenum(VI) dioxide bis(acetylacetonate), Molybdenyl acetylacetonate, Bis(acetylacetonato)dioxomolybdenum(VI)

M0464-25G M0464-5G
TCIAM0464-25GEA 214 EUR
TCIAM0464-25G TCIAM0464-5G
Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥95.0% (by titrimetric analysis)
Bis(2,4-pentanedionato)molybdenum(VI) dioxide
Kaava: C₁₀H₁₄MoO₆
MDL numero: MFCD00011506
CAS-numero: 17524-05-9
EINECS: 241-522-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e598d64b7ef8acadb7d67ae31c3cdeb254f03930&vid=e598d64b7ef8acadb7d67ae31c3cdeb254f03930&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light yellow to Brown powder to crystal
Purity (Chelometric Titration) min. 95.0 %
Solubility in hot EtOH very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR