Methyl cellulose, 13-18 mPa.s (20 °C) (C=2%, H₂O)

Toimittaja: TCI

Synonyms: Cellulose methyl ether

M0290-500G M0290-25G
TCIAM0290-500GEA 71.5 EUR
TCIAM0290-500G TCIAM0290-25G
Methyl cellulose, 13-18 mPa.s (20 °C) (C=2%, H₂O)
Methyl cellulose
MDL numero: MFCD00081763
CAS-numero: 9004-67-5
Merck Index: 14,06040

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Drying loss max. 8.0 %
Ignition residue (Sulfate) max. 1.5 %
Viscosity 13.0 to 18.0 mPa-s (2 %, H2O, 20 deg-C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR