2,2'-Methylenebis(6-tert-butyl-p-cresol) ≥99.0% (by GC)

Toimittaja: TCI
Varoitus

M0217-25G M0217-250G
TCIAM0217-25GEA 19.8 EUR
TCIAM0217-25G TCIAM0217-250G
2,2'-Methylenebis(6-tert-butyl-p-cresol) ≥99.0% (by GC)
2,2'-Methylenebis(6-tert-butyl-p-cresol)
Kaava: C₂₃H₃₂O₂
MDL numero: MFCD00043641
CAS-numero: 119-47-1
EINECS: 204-327-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d1e7c4a0c9ce6a9c7435f2c43220d12e7cd8e3b&vid=0d1e7c4a0c9ce6a9c7435f2c43220d12e7cd8e3b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 130.0 to 134.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR