(±)-2-Metyylivoihappo ≥97.0% (by GC)

Toimittaja: TCI
Vaara

M0181-25ML M0181-500ML
TCIAM0181-25MLEA 24.2 EUR
TCIAM0181-25ML TCIAM0181-500ML
(±)-2-Metyylivoihappo ≥97.0% (by GC)
(±)-2-Metyylivoihappo
Kaava: C₅H₁₀O₂
Kiehumispiste: 174°C
Sulamispiste: –70 °C
Tiheys: 0,938 g/cm³ (20 °C)
Leimahduspiste: 73°C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002669
CAS-numero: 116-53-0
EINECS: 204-145-2
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39989b049c18333c8b8c3fa56ece63f47a08a300&vid=39989b049c18333c8b8c3fa56ece63f47a08a300&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR