D(+)-mannoosi ≥98.0% (by HPLC)

Toimittaja: TCI

M0045-500G M0045-25G M0045-100G
TCIAM0045-500GEA 347 EUR
TCIAM0045-500G TCIAM0045-25G TCIAM0045-100G
D(+)-mannoosi ≥98.0% (by HPLC)
D(+)-mannoosi
Kaava: C₆H₁₂O₆
Kiehumispiste: 376 °C (1013 hPa)
Sulamispiste: 133 °C
Tiheys: 1,539 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064122
CAS-numero: 3458-28-4
EINECS: 222-392-4
Merck Index: 14,05747
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f13f3d32c18ff7ed2ef2f0afd5a9bbd0fbcf5a16&vid=f13f3d32c18ff7ed2ef2f0afd5a9bbd0fbcf5a16&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D +14.0 to +15.5 deg (C=4, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR