Levocetirizine dihydrochloride ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

L0264-1G L0264-5G
TCIAL0264-1GEA 33 EUR
TCIAL0264-1G TCIAL0264-5G
Levocetirizine dihydrochloride ≥98.0% (by HPLC, titration analysis)
Levocetirizine dihydrochloride
Kaava: C₂₁H₂₅ClN₂O₃
MDL numero: MFCD07366507
CAS-numero: 130018-87-0
Merck Index: 14,02022
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d3555bcc91d96c62ebd090007c4df0d9d56e860&vid=3d3555bcc91d96c62ebd090007c4df0d9d56e860&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR