Lithium trifluoro(trifluoromethyl)borate - dimethyl carbonate complex ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

L0240-1G L0240-5G
TCIAL0240-1GEA 168 EUR
TCIAL0240-1G TCIAL0240-5G
Lithium trifluoro(trifluoromethyl)borate - dimethyl carbonate complex ≥98.0% (by titrimetric analysis)
Lithium trifluoro(trifluoromethyl)borate - dimethyl carbonate complex
Kaava: CBF₆Li
Storage Temperature: Pakastin
CAS-numero: 1443685-69-5

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR