Loperamide hydrochloride ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Vaara

L0154-25G L0154-5G
TCIAL0154-25GEA 368 EUR
TCIAL0154-25G TCIAL0154-5G
Loperamide hydrochloride ≥98.0% (by HPLC, titration analysis)
Loperamide hydrochloride
Kaava: C₂₉H₃₃ClN₂O₂
Sulamispiste: 223°C
MDL numero: MFCD00058581
CAS-numero: 34552-83-5
EINECS: 252-082-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,05571

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR