Linolihappo ≥97.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid

L0124-25G L0124-5G
TCIAL0124-25GEA 198 EUR
TCIAL0124-25G TCIAL0124-5G
Linolihappo ≥97.0% (by GC, titration analysis)
Linolihappo
Kaava: C₁₈H₃₂O₂
Kiehumispiste: 177°C
Sulamispiste: –5°C
Tiheys: 0,9 g/cm³ (20 °C)
Storage Temperature: Jääkaappi
MDL numero: MFCD00064241
CAS-numero: 60-33-3
EINECS: 200-470-9
Merck Index: 14,05505
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf291b412effb9342c01769ad8e0b142940020f8&vid=bf291b412effb9342c01769ad8e0b142940020f8&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR