Lactobionic acid ≥96.0% (by titrimetric analysis) (mixture of Acid form and Lactone form)

Toimittaja: TCI

L0005-25G L0005-500G L0005-100G
TCIAL0005-25GEA 53.9 EUR
TCIAL0005-25G TCIAL0005-500G TCIAL0005-100G
Lactobionic acid ≥96.0% (by titrimetric analysis) (mixture of Acid form and Lactone form)
Lactobionic acid
Kaava: C₁₂H₂₂O₁₂
MDL numero: MFCD00078147
CAS-numero: 96-82-2
EINECS: 202-538-3
Merck Index: 14,05342
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58a1136f93ca8dc379a41fa74979164397fc270e&vid=58a1136f93ca8dc379a41fa74979164397fc270e&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 96.0 to 106.0 %
Specific rotation [a]20/D +21.0 to +27.0 deg (C=8.8, H2O) (after 4hrs)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR