K02288 ≥98.0% (by HPLC)

Toimittaja: TCI
Varoitus

K0065-10MG K0065-50MG
TCIAK0065-10MGEA 114 EUR
TCIAK0065-10MG TCIAK0065-50MG
K02288 ≥98.0% (by HPLC)
K02288
Kaava: C₂₀H₂₀N₂O₄
Storage Temperature: Jääkaappi
MDL numero: MFCD26936347
CAS-numero: 1431985-92-0

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 7.50 to 8.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR