2-Jodi-5-nitrotolueeni ≥96.0% (by GC)

Toimittaja: TCI
Varoitus

I0992-5G I0992-25G I0992-1G
TCIAI0992-5GEA 120 EUR
TCIAI0992-5G TCIAI0992-25G TCIAI0992-1G
2-Jodi-5-nitrotolueeni ≥96.0% (by GC)
2-Jodi-5-nitrotolueeni
Kaava: C₇H₆INO₂
MDL numero: MFCD00051517
CAS-numero: 5326-38-5
EINECS: 226-204-1

Order Now

Testitulosten erittely

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 96.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR