Iloperidone ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

I0926-25MG I0926-100MG I0926-1G
TCIAI0926-25MGEA 18.7 EUR
TCIAI0926-25MG TCIAI0926-100MG TCIAI0926-1G
Iloperidone ≥98.0% (by HPLC, titration analysis)
Iloperidone
Kaava: C₂₄H₂₇FN₂O₄
Storage Temperature: Pakastin
MDL numero: MFCD00866688
CAS-numero: 133454-47-4
Merck Index: 14,04900
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dbb60328bd0af01717629e3d530d8e387bcd295&vid=6dbb60328bd0af01717629e3d530d8e387bcd295&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR