Ipratropium bromide monohydrate ≥98.0% (by HPLC, total nitrogen)

Toimittaja: TCI
Varoitus

I0907-200MG I0907-1G
TCIAI0907-200MGEA 84.7 EUR
TCIAI0907-200MG TCIAI0907-1G
Ipratropium bromide monohydrate ≥98.0% (by HPLC, total nitrogen)
Ipratropium bromide monohydrate
Kaava: C₂₀H₃₀BrNO₃
MDL numero: MFCD09842549
CAS-numero: 66985-17-9

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on anh.substance)
Water 3.5 to 4.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR