Imidapril hydrochloride ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

I0901-25MG I0901-100MG
TCIAI0901-25MGEA 107 EUR
TCIAI0901-25MG TCIAI0901-100MG
Imidapril hydrochloride ≥98.0% (by HPLC, titration analysis)
Imidapril hydrochloride
Kaava: C₂₀H₂₇N₃O₆
MDL numero: MFCD00923828
CAS-numero: 89396-94-1
Merck Index: 14,04911
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b6bc73e95054c53d00652a7b3b75f6ee4f21826&vid=3b6bc73e95054c53d00652a7b3b75f6ee4f21826&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -65.0 to -69.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR