Rauta(II)asetaatti ≥90.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

I0765-25G I0765-5G
TCIAI0765-25GEA 153 EUR
TCIAI0765-25G TCIAI0765-5G
Rauta(II)asetaatti ≥90.0% (by titrimetric analysis)
Rauta(II)asetaatti
Kaava: C₄H₆FeO₄
Sulamispiste: 190 °C
MDL numero: MFCD00058909
CAS-numero: 3094-87-9
EINECS: 221-441-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65f240c67037c09d18e418d10e1b87d8d5de4a5a&vid=65f240c67037c09d18e418d10e1b87d8d5de4a5a&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Gray to Brown powder to crystal
Purity (Potassium permanganate method) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR