Jodi ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

I0604-25G I0604-500G
TCIAI0604-25GEA 24.2 EUR
TCIAI0604-25G TCIAI0604-500G
Jodi ≥98.0% (by titrimetric analysis)
Jodi
Kaava: I₂
Kiehumispiste: 184,3 °C (1013 hPa)
Sulamispiste: 114°C
Tiheys: 4,9 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011355
CAS-numero: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8,III
Merck Index: 14,05014
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ad8a29196f0b4f7e8bc4b2c5a31d4ccae809a81&vid=5647cb2397f853e7bdf732d4e548f0313a5a2ca2&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Gray to Dark blue to Black powder to crystal
Purity (Iodometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR