3,5-Bis(trifluorimetyyli)fenyyli-isotiosyanaatti ≥98.0% (by GC)

Toimittaja: TCI
Vaara

I0569-25G I0569-5G
TCIAI0569-25GEA 274 EUR
TCIAI0569-25G TCIAI0569-5G
3,5-Bis(trifluorimetyyli)fenyyli-isotiosyanaatti ≥98.0% (by GC)
3,5-Bis(trifluorimetyyli)fenyyli-isotiosyanaatti
Kaava: C₉H₃F₆NS
Kiehumispiste: 95°C
Tiheys: 1,48 g/cm³ (20 °C)
Leimahduspiste: 108°C
MDL numero: MFCD00040838
CAS-numero: 23165-29-9
UN: 1760
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd190923bfc37b79af012dc15a65585a2e7210e7&vid=bd190923bfc37b79af012dc15a65585a2e7210e7&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR