2-Propanoli ≥99.5% (by GC) for spectrophotometry

Toimittaja: TCI
Vaara

Synonyms: Isopropyylialkoholi

I0164-100ML I0164-500ML
TCIAI0164-100MLEA 17.6 EUR
TCIAI0164-100ML TCIAI0164-500ML
2-Propanoli ≥99.5% (by GC) for spectrophotometry
2-Propanoli
Kaava: (CH₃)₂CHOH
Kiehumispiste: 82°C
Sulamispiste: –89 °C
Tiheys: 0,786 g/cm³ (20 °C)
Leimahduspiste: 15°C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011674
CAS-numero: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 14,05208
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ef0a3050bc3fd54fa52c35e004090bd986d097f&vid=695ce94126a68bd2a2e0e9965b4f910241212a78&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless clear liquid
Purity (GC) min. 99.5 %
Absorbance (210nm) max. 0.800
Absorbance (220nm) max. 0.400
Absorbance (230nm) max. 0.200


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR