3-(Indol-3-yyli)propaanihappo ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

I0032-25G I0032-5G
TCIAI0032-25GEA 80.3 EUR
TCIAI0032-25G TCIAI0032-5G
3-(Indol-3-yyli)propaanihappo ≥98.0% (by titrimetric analysis)
3-(Indol-3-yyli)propaanihappo
Kaava: C₁₁H₁₁NO₂
Sulamispiste: 129…132 °C
Tiheys: 1,356 g/cm³ (20 °C)
MDL numero: MFCD00005660
CAS-numero: 830-96-6
EINECS: 212-600-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e6e1ff00284f809045e9bfbf9d95cc93cf3e68e&vid=1e6e1ff00284f809045e9bfbf9d95cc93cf3e68e&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 132.0 to 136.0 deg-C
Solubility in Methanol very faint turbidity

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR