(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥97.0% (by titrimetric analysis)

Toimittaja: TCI

Synonyms: H-HoArg-OH·HCl, L(+)-Homoarginine hydrochloride, H-HomoArg-OH.HCl, H-HomoArg-OH·HCl

H1172-1G H1172-5G
TCIAH1172-1GEA 57.2 EUR
TCIAH1172-1G TCIAH1172-5G
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride ≥97.0% (by titrimetric analysis)
(S)-2-Amino-6-guanidinohexanoic acid hydrochloride
Kaava: C₇H₁₆N₄O₂·HCl
MW: 224.7 g/mol
Sulamispiste: 206°C
MDL numero: MFCD00012622
CAS-numero: 1483-01-8
EINECS: 216-045-6

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Specific rotation [a]20/D +16.0 to +20.0 deg (C=8,6N HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR