4-(3-Hydroxyphenyl)piperidine ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

H0965-25G H0965-5G
TCIAH0965-25GEA 2010 EUR
TCIAH0965-25G TCIAH0965-5G
4-(3-Hydroxyphenyl)piperidine ≥98.0% (by GC)
4-(3-Hydroxyphenyl)piperidine
Kaava: C₁₁H₁₅NO
MDL numero: MFCD02093474
CAS-numero: 110878-71-2

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR