Methyl-5-(hydroxymethyl)-1H-imidazole-4-carboxylate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

H0840-1G H0840-5G
TCIAH0840-1GEA 249 EUR
TCIAH0840-1G TCIAH0840-5G
Methyl-5-(hydroxymethyl)-1H-imidazole-4-carboxylate ≥98.0% (by titrimetric analysis)
Methyl-5-(hydroxymethyl)-1H-imidazole-4-carboxylate
Kaava: C₆H₈N₂O₃
MDL numero: MFCD00191520
CAS-numero: 82032-43-7

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR