Hexylene di(acetate) ≥98.0% (by GC)

Toimittaja: TCI

H0832-25ML H0832-500ML
TCIAH0832-25MLEA 17.6 EUR
TCIAH0832-25ML TCIAH0832-500ML
Hexylene di(acetate) ≥98.0% (by GC)
Hexylene di(acetate)
Kaava: C₁₀H₁₈O₄
Kiehumispiste: 260°C
MDL numero: MFCD00191655
CAS-numero: 6222-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1777e51cffbeb555ede734aa569e5fce67710591&vid=1777e51cffbeb555ede734aa569e5fce67710591&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR