2-Hydroxy-2-methylbutyric acid ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

H0636-25G H0636-5G
TCIAH0636-25GEA 131 EUR
TCIAH0636-25G TCIAH0636-5G
2-Hydroxy-2-methylbutyric acid ≥98.0% (by titrimetric analysis)
2-Hydroxy-2-methylbutyric acid
Kaava: C₅H₁₀O₃
Kiehumispiste: 109°C
MDL numero: MFCD00004476
CAS-numero: 3739-30-8
EINECS: 223-120-7

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 69.0 to 74.0 deg-C
Solubility in Water within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR