Hexyltrimethylammonium bromide ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

H0534-25G H0534-500G
TCIAH0534-25GEA 75.9 EUR
TCIAH0534-25G TCIAH0534-500G
Hexyltrimethylammonium bromide ≥98.0% (by HPLC, titration analysis)
Hexyltrimethylammonium bromide
Kaava: C₉H₂₂BrN
MDL numero: MFCD00051872
CAS-numero: 2650-53-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd1cfd60462a81009e030197ed1638ddc49c3ab4&vid=fd1cfd60462a81009e030197ed1638ddc49c3ab4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 183.0 to 187.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR