Heptafluorobutyryl chloride ≥95.0% (by GC) for heptafluorobutyration

Toimittaja: TCI
Vaara

H0508-25G H0508-5G
TCIAH0508-25GEA 107 EUR
TCIAH0508-25G TCIAH0508-5G
Heptafluorobutyryl chloride ≥95.0% (by GC) for heptafluorobutyration
Kaava: C₄ClF₇O
Kiehumispiste: 39°C
Tiheys: 1,55 g/cm³ (25 °C)
Storage Temperature: Jääkaappi
MDL numero: MFCD00000707
CAS-numero: 375-16-6
EINECS: 206-785-8
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5a6a73240ca4528cbda7cfc5eee8037b76ef018&vid=cfec4481e8f1f886f7b38c0f83db98ecc6647056&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR