Asetoiini ≥98.0% (by GC) (May exist as crystalline dimer)

Toimittaja: TCI
Varoitus

H0225-25G H0225-500G
TCIAH0225-25GEA 27.5 EUR
TCIAH0225-25G TCIAH0225-500G
Asetoiini ≥98.0% (by GC) (May exist as crystalline dimer)
Asetoiini
Kaava: C₄H₈O₂
Kiehumispiste: 144°C
Sulamispiste: 20 °C
Tiheys: 0,998 g/cm³ (20 °C)
Leimahduspiste: 46°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00004521
CAS-numero: 513-86-0
EINECS: 208-174-1
UN: 2621
ADR: 3,III
Merck Index: 14,00064
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ec11d2db1ea00b7885a9d51a04d2f25285ae314&vid=e9029a8ae14be8319ff9e0896ff914d6328edeb8&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR