L(+)-Histidiinimonohydrokloridimonohydraatii ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

Synonyms: L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate

H0150-25G H0150-500G H0150-100G
TCIAH0150-25GEA 17.6 EUR
TCIAH0150-25G TCIAH0150-500G TCIAH0150-100G
L(+)-Histidiinimonohydrokloridimonohydraatii ≥98.0% (by HPLC, titration analysis)
L(+)-Histidiinimonohydrokloridimonohydraatii
Kaava: C₆H₉N₃O₂·HCl·H₂O
Sulamispiste: 254°C
Tiheys: 1,485 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00151027
CAS-numero: 5934-29-2
EINECS: 211-438-9
Merck Index: 14,04720
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e7c134209feb67ac97eaff427b6f92f4f72ca8e&vid=6e7c134209feb67ac97eaff427b6f92f4f72ca8e&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +8.9 to +9.6 deg (C=8, HCl (1+1))


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR