Hexaphenyldisilane ≥97.0% (by GC)

Toimittaja: TCI
Varoitus

H0119-1G H0119-5G
TCIAH0119-1GEA 51.7 EUR
TCIAH0119-1G TCIAH0119-5G
Hexaphenyldisilane ≥97.0% (by GC)
Hexaphenyldisilane
Kaava: C₃₆H₃₀Si₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00039558
CAS-numero: 1450-23-3
EINECS: 215-913-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efc189e8a4a788ffb24b95256eaad021b9e031e2&vid=efc189e8a4a788ffb24b95256eaad021b9e031e2&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Solubility in hot Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR