Monoethyl glutarate ≥98.0% (by GC, titration analysis)

Toimittaja: TCI

G0271-25G G0271-5G
TCIAG0271-25GEA 97.9 EUR
TCIAG0271-25G TCIAG0271-5G
Monoethyl glutarate ≥98.0% (by GC, titration analysis)
Monoethyl glutarate
Kaava: C₇H₁₂O₄
Kiehumispiste: 170°C
MDL numero: MFCD00014379
CAS-numero: 1070-62-8
EINECS: 213-977-5

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR