Glycyl-l-proline
Toimittaja: TCI
G0137-1G
TCIAG0137-1GEA 146 EUR
TCIAG0137-1G
Glycyl-l-proline
Proteiinit
  • Specific rotation [a]20/D: –114° (C=4, H₂O)
  • Solubility in water: very faint turbidity
Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Pro-OH