Nalpha-Glycyl-L-asparagine

Toimittaja: TCI

G0118-1G
TCIAG0118-1GEA 152 EUR
TCIAG0118-1G
Nalpha-Glycyl-L-asparagine
Proteiinit

  • Specific rotation [a]20/D: –8° (C=1, H₂O)
  • Solubility in water: almost transparency
  • EINECS: 217-881-4


Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: H-Gly-Asn-OH

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR