L(+)-glutamiinihappo ≥99.0% (by titrimetric analysis)

Toimittaja: TCI

G0059-25G G0059-500G
TCIAG0059-25GEA 19.8 EUR
TCIAG0059-25G TCIAG0059-500G
L(+)-glutamiinihappo ≥99.0% (by titrimetric analysis)
L(+)-glutamiinihappo
Kaava: HO₂CCH₂CH₂CH(NH₂)CO₂H
Kiehumispiste: 325 °C (1013 hPa)
Sulamispiste: 160 °C
Tiheys: 1,538 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002634
CAS-numero: 56-86-0
EINECS: 200-293-7
Merck Index: 14,04469
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=455d402740b3250b1bdaa1629d94ba3fffeb786d&vid=455d402740b3250b1bdaa1629d94ba3fffeb786d&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D +31.5 to +32.5 deg (C=5, HCl (1+5))
Solubility in dilute HCl almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR