D-(+)-Glucosamine hydrochloride ≥98.0% (by titrimetric analysis)

Toimittaja: TCI

G0044-25G G0044-500G
TCIAG0044-25GEA 23.1 EUR
TCIAG0044-25G TCIAG0044-500G
D-(+)-Glucosamine hydrochloride ≥98.0% (by titrimetric analysis)
D-(+)-Glucosamine hydrochloride
Kaava: C₆H₁₃NO₅
MDL numero: MFCD00135831
CAS-numero: 66-84-2
EINECS: 200-638-1

Order Now

Testitulosten erittely

Appearance White powder to crystaline
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D +70.0 to +74.0 deg (C=1, H2O) (after 20hours)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR