Fargesin ≥98.0% (by HPLC)

Toimittaja: TCI

F1188-10MG F1188-50MG
TCIAF1188-10MGEA 231 EUR
TCIAF1188-10MG TCIAF1188-50MG
Fargesin ≥98.0% (by HPLC)
Fargesin
Kaava: C₂₁H₂₂O₆
MDL numero: MFCD07781423
CAS-numero: 31008-19-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=524496f8de72b00d43add3c1580d5ca394b7c1d5&vid=524496f8de72b00d43add3c1580d5ca394b7c1d5&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 132.0 to 136.0 deg-C
Specific rotation [a]20/D +113 to +125 deg (C=1, CH2CL2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR