4'-Fluoro-3'-methylacetophenone ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

F1097-1G F1097-5G
TCIAF1097-1GEA 28.6 EUR
TCIAF1097-1G TCIAF1097-5G
4'-Fluoro-3'-methylacetophenone ≥98.0% (by GC)
4'-Fluoro-3'-methylacetophenone
Kaava: C₉H₉FO
Kiehumispiste: 215°C
MDL numero: MFCD04972879
CAS-numero: 369-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d00b2d8c524e1885b3a331ad7718bd0f08cdc90d&vid=d00b2d8c524e1885b3a331ad7718bd0f08cdc90d&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR