Formamidine hydrobromide, low water content ≥98.0% (by total nitrogen and titration analysis)

Toimittaja: TCI

F0973-1G F0973-5G F0973-25G
TCIAF0973-1GEA 51.7 EUR
TCIAF0973-1G TCIAF0973-5G TCIAF0973-25G
Formamidine hydrobromide, low water content ≥98.0% (by total nitrogen and titration analysis)
Formamidine hydrobromide
Kaava: CH₄N₂·HBr
MDL numero: MFCD28369274
CAS-numero: 146958-06-7

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR