8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid ≥97.0% (by HPLC)

Toimittaja: TCI

F0719-1G F0719-200MG
TCIAF0719-1GEA 248 EUR
TCIAF0719-1G TCIAF0719-200MG
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid ≥97.0% (by HPLC)
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid
Kaava: C₂₁H₂₃NO₆
MDL numero: MFCD01321015
CAS-numero: 166108-71-0

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR