4-Fluori-2-metoksibentsoehappo ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

F0680-1G F0680-5G
TCIAF0680-1GEA 29.7 EUR
TCIAF0680-1G TCIAF0680-5G
4-Fluori-2-metoksibentsoehappo ≥98.0% (by GC, titration analysis)
4-Fluori-2-metoksibentsoehappo
Kaava: C₈H₇FO₃
MDL numero: MFCD00673010
CAS-numero: 395-82-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=609f1c46fbe8da300ba7bcfa8e956b695f65643a&vid=609f1c46fbe8da300ba7bcfa8e956b695f65643a&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 135.0 to 139.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR