1,1'-Ferrocenediboronic acid ≥95.0% (by titrimetric analysis) (contains varying amounts of Anhydride)

Toimittaja: TCI
Varoitus

F0664-1G
TCIAF0664-1GEA 101 EUR
TCIAF0664-1G
1,1'-Ferrocenediboronic acid ≥95.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
1,1'-Ferrocenediboronic acid
Kaava: C₁₀H₁₂B₂FeO₄
MDL numero: MFCD00799542
CAS-numero: 32841-83-1

Order Now

Testitulosten erittely

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (Chelometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR