2-Fluoro-α-methyl-4-biphenylacetic acid ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Vaara

F0371-25G F0371-5G
TCIAF0371-25GEA 264 EUR
TCIAF0371-25G TCIAF0371-5G
2-Fluoro-α-methyl-4-biphenylacetic acid ≥98.0% (by HPLC, titration analysis)
2-Fluoro-α-methyl-4-biphenylacetic acid
Kaava: C₁₅H₁₃FO₂
MDL numero: MFCD00079303
CAS-numero: 5104-49-4
EINECS: 225-827-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,04199
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e95cf4bcaf6d3a3f65d50a46353394e13eeae36&vid=2e95cf4bcaf6d3a3f65d50a46353394e13eeae36&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 116.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR