N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥95.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

F0358-25G F0358-5G F0358-100G
TCIAF0358-25GEA 94.6 EUR
TCIAF0358-25G TCIAF0358-5G TCIAF0358-100G
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate) ≥95.0% (by titrimetric analysis)
N-Fluoro-N'-(chloromethyl)triethylenediaminebis(tetrafluoroborate)
Kaava: C₇H₁₄B₂ClF₉N₂
MDL numero: MFCD00142607
CAS-numero: 140681-55-6
EINECS: 414-380-4
UN: 3088
ADR: 4.2,II
Merck Index: 14,08425

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (Iodometric Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR