Propyl-2-furancarboxylate ≥97.0% (by GC)

Toimittaja: TCI

F0155-25ML
TCIAF0155-25MLEA 75.9 EUR
TCIAF0155-25ML
Propyl-2-furancarboxylate ≥97.0% (by GC)
Propyl-2-furancarboxylate
Kaava: C₈H₁₀O₃
Kiehumispiste: 211°C
MDL numero: MFCD00046108
CAS-numero: 615-10-1
EINECS: 210-407-7

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR