Ethyl fluoroacetate ≥98.0% (by GC)

Toimittaja: TCI
Vaara

F0030-25ML F0030-500ML F0030-100ML
TCIAF0030-25MLEA 33 EUR
TCIAF0030-25ML TCIAF0030-500ML TCIAF0030-100ML
Ethyl fluoroacetate ≥98.0% (by GC)
Ethyl fluoroacetate
Kaava: C₄H₇FO₂
Kiehumispiste: 117°C
Tiheys: 1,0912 g/cm³ (20 °C)
Leimahduspiste: 30°C
MDL numero: MFCD00000450
CAS-numero: 459-72-3
EINECS: 207-297-8
UN: 2929
ADR: 6.1,II
Merck Index: 14,04167
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a60f6b18b1a62b3e1358483c4370963ecd6bc70&vid=1a60f6b18b1a62b3e1358483c4370963ecd6bc70&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR