Epothilone B ≥95.0% (by HPLC)

Toimittaja: TCI

E1378-25MG E1378-5MG
TCIAE1378-25MGEA 424 EUR
TCIAE1378-25MG TCIAE1378-5MG
Epothilone B ≥95.0% (by HPLC)
Epothilone B
Kaava: C₂₇H₄₁NO₆S
Sulamispiste: 94°C
Storage Temperature: Pakastin
MDL numero: MFCD02101921
CAS-numero: 152044-54-7
Merck Index: 14,03628

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Specific rotation [a]20/D -40.0 to -44.0 deg (C=0.2, methanol)
Elemental analysis (Carbon) 62.50 to 65.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR