Ethyl-2,6-dichloropyridine-3-carboxylate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

E1194-1G E1194-5G
TCIAE1194-1GEA 58.3 EUR
TCIAE1194-1G TCIAE1194-5G
Ethyl-2,6-dichloropyridine-3-carboxylate ≥98.0% (by GC)
Ethyl-2,6-dichloropyridine-3-carboxylate
Kaava: C₈H₇Cl₂NO₂
MDL numero: MFCD09757510
CAS-numero: 58584-86-4

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR