Ethyl-3-(benzylamino)-3-oxopropionate ≥98.0% (by GC)

Toimittaja: TCI

E1098-1G E1098-5G
TCIAE1098-1GEA 116 EUR
TCIAE1098-1G TCIAE1098-5G
Ethyl-3-(benzylamino)-3-oxopropionate ≥98.0% (by GC)
Ethyl-3-(benzylamino)-3-oxopropionate
Kaava: C₁₂H₁₅NO₃
MDL numero: MFCD28128094
CAS-numero: 29689-63-2

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 53.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR